missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DSCAM Monoclonal Antibody (399212)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524134
This item is not returnable.
View return policy
Description
In direct ELISAs and Western blots, no cross-reactivity with recombinant human DSCAM-L1 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
This gene is a member of the immunoglobulin superfamily of cell adhesion molecules (Ig-CAMs), and is involved in human central and peripheral nervous system development. This gene is a candidate for Down syndrome and congenital heart disease (DSCHD). A gene encoding a similar Ig-CAM protein is located on chromosome 11. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. [provided by RefSeq, Oct 2012].
Specifications
| DSCAM | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| O60469 | |
| DSCAM | |
| Mouse myeloma cell line NS0-derived recombinant human DSCAM Long Isoform Glu18-Met1595. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot, Immunocytochemistry | |
| 399212 | |
| Unconjugated | |
| DSCAM | |
| 4932410A21Rik; CHD2; CHD2-42; CHD2-52; Down syndrome cell adhesion molecule; Down syndrome cell adhesion molecule homolog; DS cell adhesion molecule; Dscam; LOW QUALITY PROTEIN: Down syndrome cell adhesion molecule | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 1826 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction