missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DSCC1 Antibody (V76P1B2*B12), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50380B
This item is not returnable.
View return policy
Description
DSCC1 Monoclonal specifically detects DSCC1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| DSCC1 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| DSCC1 | |
| Peptide Sequence - KVYNSRRPIS (immunogen peptide is identical in sheep, bovine, canine, equine, chicken and zebrafish) | |
| 0.1 mL | |
| Epigenetics | |
| 79075 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry | |
| V76P1B2*B12 | |
| Western Blot, ELISA, Immunohistochemistry | |
| DCC1MGC5528, defective in sister chromatid cohesion 1 homolog (S. cerevisiae), Defective in sister chromatid cohesion protein 1 homolog, hDCC1, sister chromatid cohesion protein DCC1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction