Learn More
Invitrogen™ DSCC1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA599006
Product Code. 16501832
  • € 464.00 / 100µg
Description
DSCC1 Polyclonal Antibody for Western Blot, ELISA
CHTF18 (MIM 613201), CHTF8 (MIM 613202), and DSCC1 arecomponents of an alternative replication factor C (RFC) (see MIM600404) complex that loads PCNA (MIM 176740) onto DNA during Sphase of the cell cycle.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DSCC1
Polyclonal
Unconjugated
DSCC1
2010006I05Rik; 2600005O03Rik; DCC1; defective in sister chromatid cohesion 1 homolog; defective in sister chromatid cohesion 1 homolog (S. cerevisiae); defective in sister chromatid cohesion protein 1 homolog; DNA replication and sister chromatid cohesion 1; DSCC1; RGD1561749; Sister chromatid cohesion protein DCC1; UNQ9337/PRO34008
Rabbit
Protein G
RUO
79075
-20°C or -80°C if preferred
Liquid
ELISA, Western Blot
4 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Q9BVC3
DSCC1
Recombinant Human Sister chromatid cohesion protein DCC1 protein (1-393AA).