missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: FRKTAERLLR FSDKRQTEEA IDRIFAISQQ QQQQSKSKKN RRAGKRYKFV DAVPDIFAQI EVNEKKIRQK AQILAQKELP IDEDEEMKDL LDFADVTYEK HKNGGLIKGR FGQARMVTTT HSRAPSLSAS YT Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 89%.
Converts D-glucuronic acid to L-iduronic acid (IdoUA) residues.
Specifications
Specifications
| Antigen | DSE |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | DSE |
| Gene Accession No. | Q9UL01 |
| Gene Alias | 6030499O08; AI480506; B130024B19Rik; Chondroitin-glucuronate 5-epimerase; dermatan sulfate epimerase; dermatan-sulfate epimerase; DS epimerase; Dse; DSEP; DSEPI; DS-epi1; EDSMC2; Sart2; SART-2; squamous cell carcinoma antigen recognized by T cells 2; squamous cell carcinoma antigen recognized by T-cells 2 |
| Gene Symbols | DSE |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
