missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DSG2 Mouse anti-Human, Clone: 6D8, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB14502.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against full length recombinant human DSG2.
- High specificity
- High reactivity
Specifications
| DSG2 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| Q14126 | |
| DSG2 | |
| Recombinant protein corresponding to full length human DSG2. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Western Blot | |
| 6D8 | |
| Mouse monoclonal antibody raised against full length recombinant human DSG2. | |
| In 10 mM PBS (0.05% BSA, 0.05% sodium azide). | |
| ARVC10/ARVD10/CDHF5/HDGC/MGC117034/MGC117036/MGC117037 | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 1829 | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction