missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DSPG3/EPYC Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-00204CL1
This item is not returnable.
View return policy
Description
DSPG3/EPYC Polyclonal antibody specifically detects DSPG3/EPYC in Human samples. It is validated for ELISA
Specifications
| DSPG3/EPYC | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 1833 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA | |
| CoraFluor 1 | |
| Dermatan sulfate proteoglycan 3dermatan sulphate proteoglycan 3, DSPG3proteoglycan-lb, epiphycan, Pg-Lb, PGLB, Proteoglycan-Lb, SLRR3Bepiphycan proteoglycan, Small chondroitin/dermatan sulfate proteoglycan | |
| Produced in rabbits immunized with purified, recombinant Human DSPG3/EPYC (Accession#: AAH30958.1; Met1-Val322) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction