missing translation for 'onlineSavingsMsg'
Learn More
Learn More
dsRNA Antibody (1D3), DyLight 650 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20111C
This item is not returnable.
View return policy
Description
dsRNA Monoclonal antibody specifically detects dsRNA in Broad Species Reactivity samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| dsRNA | |
| Monoclonal | |
| DyLight 650 | |
| Double-stranded RNA | |
| RDV-RNA-methylated bovine serum albumin complex. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG κ |
| Western Blot, ELISA, Immunohistochemistry | |
| 1D3 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| All species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction