missing translation for 'onlineSavingsMsg'
Learn More
Learn More
dsRNA Antibody (J5), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11396UV
This item is not returnable.
View return policy
Description
dsRNA Monoclonal antibody specifically detects dsRNA in Broad Species Reactivity samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunoblot
Specifications
| dsRNA | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Broad Species Reactivity | |
| Purified |
| ELISA, Immunohistochemistry, Immunofluorescence, Immunoblot | |
| J5 | |
| ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoblotting | |
| Double-stranded RNA | |
| Female DBA/2 mice were injected intraperitonially with a mixture of 50 ug L-dsRNA and 75 ug methylated bovine serum albumin, emulsified in complete Freund's adjuvant. After several boosts spleen cells were fused with Sp2/0-Agl4 myeloma cells to generate the hybridoma clone. | |
| 0.2 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction