Learn More
dsRNA Antibody (J5) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-11396JF525
Product Code. 30175244
  • ONLINE EXCLUSIVE PRICE
    € 890.00
    € 841.05 / 0.20mL
    Save € 48.95
    5% Off
Description
dsRNA Monoclonal antibody specifically detects dsRNA in Broad Species Reactivity samples. It is validated for ELISA,Immunohistochemistry,Immunoblot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
dsRNA
Monoclonal
Janelia Fluor 525
Double-stranded RNA
Female DBA/2 mice were injected intraperitonially with a mixture of 50 μg L-dsRNA and 75 μg methylated bovine serum albumin, emulsified in complete Freund's adjuvant. After several boosts spleen cells were fused with Sp2/0-Agl4 myeloma cells to generate the hybridoma clone.
0.2 mL
Primary
Store at 4°C in the dark.
IgG2b κ
ELISA, Immunohistochemistry, Immunoblot, Immunocytochemistry/Immunofluorescence
J5
50mM Sodium Borate
Mouse
Protein A purified
RUO
Broad Species Reactivity