missing translation for 'onlineSavingsMsg'
Learn More

dsRNA Mouse anti-Broad Species Reactivity, Clone: J5, Novus Biologicals™

Mouse Monoclonal Antibody

€ 772.00 - € 1185.00

Specifications

Antigen dsRNA
Clone J5
Concentration LYOPH
Dilution ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoblotting
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunoblot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18391342
View Documents
Novus Biologicals
NBP3-11396-200UG
200 μg
€ 772.00
200µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18392977
View Documents
Novus Biologicals
NBP3-11396-500UG
500 μg
€ 1185.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

dsRNA Monoclonal antibody specifically detects dsRNA in Broad Species Reactivity samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunoblot
Specifications

Specifications

dsRNA
LYOPH
ELISA, Immunohistochemistry, Immunofluorescence, Immunoblot
Unconjugated
Mouse
Multi-species
Double-stranded RNA
IgG2b κ
Protein A purified
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
J5
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoblotting
Monoclonal
Purified
RUO
Lyophilised from PBS
Female DBA/2 mice were injected intraperitonially with a mixture of 50 ug L-dsRNA and 75 ug methylated bovine serum albumin, emulsified in complete Freund's adjuvant. After several boosts spleen cells were fused with Sp2/0-Agl4 myeloma cells to generate the hybridoma clone.
Primary
The lyophilised sample should be reconstituted with 200 ul (500 ul for 500 ug size) sterile distilled water. The mAb will then be in PBS without any stabilisers or preservatives at a concentration of 1 mg/ml.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.