Learn More
IKA™ DT-50 Tubes with Rotor Stator Element, 50mL
Product Code. 10448283 Shop All IKA Products
Click to view available options
For Use With (Equipment):
DT-50
Unit Size:
Pack of 10
Product Code. 10448283
Brand: IKA™ 0003699600
Only null left
Add to basket
Add to basket
Use these tubes for a variety of applications. IKA™ DT-50 Tubes with Rotor Stator Element are ideal for dispersion, homogenization and suspensions.
TRUSTED_SUSTAINABILITY
Specifications
Product Type DT Tube
Volume Range (Metric) 15 to 50 mL
For Use With (Application) Dispersion, homogenisation, suspensions
For Use With (Equipment) DT-50
Includes Tube, dispersing device, cover
Packaging Quantity 10/Pk.