missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dtk/TYRO3 Antibody (109646), FITC, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB759F
This item is not returnable.
View return policy
Description
Dtk/TYRO3 Monoclonal antibody specifically detects Dtk/TYRO3 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA Standard (Matched Pair), ELISA (Matched Antibody Pair), ELISA (Matched Antibody Pair), Immunocytochemistry, CyTOF
Specifications
| Dtk/TYRO3 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair) | |
| 109646 | |
| Western Blot, Flow Cytometry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Immunocytochemistry, CyTOF-ready | |
| Brt, BYK, DTK, EC 2.7.10, EC 2.7.10.1, FLJ16467, RSE, Sky, Tif, TYRO3 protein tyrosine kinase, Tyrosine-protein kinase byk, Tyrosine-protein kinase DTK, tyrosine-protein kinase receptor TYRO3, Tyrosine-protein kinase RSE, Tyrosine-protein kinase SKY | |
| Mouse myeloma cell line NS0-derived recombinant mouse Tyro3/Dtk, Ala31-Ser418, Accession # P55144 | |
| 0.5 mL | |
| Cancer, Neuroscience, Neurotransmission, Protein Kinase, Tyrosine Kinases | |
| 7301 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction