Learn More
Dtk/TYRO3 Antibody (109646), Janelia Fluor™ 646, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB759J
Product Code. 18066755
  • € 908.00 / 0.50mL
Description
Dtk/TYRO3 Monoclonal antibody specifically detects Dtk/TYRO3 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, ELISA (Matched Antibody Pair)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Dtk/TYRO3
Monoclonal
Janelia Fluor 646
Brt, BYK, DTK, EC 2.7.10, EC 2.7.10.1, FLJ16467, RSE, Sky, Tif, TYRO3 protein tyrosine kinase, Tyrosine-protein kinase byk, Tyrosine-protein kinase DTK, tyrosine-protein kinase receptor TYRO3, Tyrosine-protein kinase RSE, Tyrosine-protein kinase SKY
Mouse myeloma cell line NS0-derived recombinant mouse Tyro3/Dtk, Ala31-Ser418, Accession # P55144
0.5 mL
Cancer, Neuroscience, Neurotransmission, Protein Kinase, Tyrosine Kinases
7301
Store at 4°C in the dark.
IgG1
Western Blot, Flow Cytometry, ELISA
109646
50mM Sodium Borate
Rat
Protein A or G purified from hybridoma culture supernatant