missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ dTRIM 24

Description
Degrader (PROTAC®) composed of the TRIM24 ligand IACS-9571 conjugated to a VHL ligand. Displays dose- and time-dependent degradation of TRIM24 in cells, with maximum degradation achieved at 5 μM. Degradation phenotypically mirrors genetic deletion of TRIM24. Demonstrates antiproliferative effects in MOLM-13 cells. Cell permeable.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| CAS | 2170695-14-2 |
| Quantity | 5 mg |
| Molecular Formula | C55H68N8O13S2 |
| Formula Weight | 1113.3 |
| Product Type | Degrader |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?