missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DTX3L Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2873AF532
This item is not returnable.
View return policy
Description
DTX3L Polyclonal antibody specifically detects DTX3L in Human samples. It is validated for Western Blot
Specifications
| DTX3L | |
| Polyclonal | |
| Western Blot | |
| BBAPEC 6.3.2.-, B-lymphoma- and BAL-associated protein, deltex 3-like (Drosophila), E3 ubiquitin-protein ligase DTX3L, Protein deltex-3-like, rhysin 2, rhysin2, Rhysin-2 | |
| The immunogen recognized by this antibody maps to a region between residue 700 and the C-terminus (residue 740) of human Deltex 3-Like Protein (B-Lymphoma- and BAL-Associated Protein) using the numbering given in entry NP_612144.1 (GeneID 151636). | |
| 0.1 mL | |
| Zinc Finger | |
| 151636 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction