Learn More
DTX3L Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB100-2873FR
Product Code. 18636769
  • € 633.00 / 0.10mL
Description
DTX3L Polyclonal antibody specifically detects DTX3L in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DTX3L
Western Blot
The immunogen recognized by this antibody maps to a region between residue 700 and the C-terminus (residue 740) of human Deltex 3-Like Protein (B-Lymphoma- and BAL-Associated Protein) using the numbering given in entry NP_612144.1 (GeneID 151636).