missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DTYMK Antibody (OTI1G8), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70604B
This item is not returnable.
View return policy
Description
DTYMK Monoclonal specifically detects DTYMK in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| DTYMK | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| DTYMK | |
| Full length recombinant protein of human DTYMK (NP_036277) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1G8 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| CDC8TMPKTYMKPP3731, deoxythymidylate kinase (thymidylate kinase), dTMP kinase, EC 2.7.4.9, FLJ44192, MGC198617, thymidylate kinase | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 1841.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction