Learn More
Dostmann Electronic Dualtemp Pro, Measuring range -33°C to 220°C
Brand:  Dostmann Electronic 5020-0413
Dualtemp Pro, Measuring range -33°C to 220°C
Product Code. 10050798
  • ONLINE EXCLUSIVE PRICE
    € 154.35
    € 89.00 / Each
    Save € 65.35
    42% Off
Only null left
Estimated Shipment: 09-02-2026