missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DUPD1 Monoclonal Antibody (OTI7E4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525336
This item is not returnable.
View return policy
Description
DUPD1 Monoclonal Antibody for Western Blot, IHC (P), Flow
Dual specificity phosphatase DUPD1 is able to dephosphorylate phosphotyrosine, phosphoserine and phosphothreonine residues, with a preference for phosphotyrosine as a substrate.
Specifications
| DUPD1 | |
| Monoclonal | |
| 1.1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q68J44 | |
| DUPD1 | |
| Full length human recombinant protein of DUPD1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI7E4 | |
| Unconjugated | |
| DUPD1 | |
| atypical dual-specific protein phosphatase; dual specificity phosphatase 27; dual specificity phosphatase and pro isomerase domain containing 1; dual specificity phosphatase DUPD1; DUPD1; DUSP27; FMDSP | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 338599 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction