Learn More
DuraCal™ Buffer pH 10.01, Hamilton™
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mL
500 mL
Description
- 10.01 pH value
- Immune to micro-organisms
- Expiration date on the bottle
- First class certificate with traceability to international standards
Specifications
Color | Yellow |
pH | 10.01 |
Physical Form | Liquid |
Quantity | 250 mL |
Solution Type | Buffer |
Solubility Information | Water Soluble |