Learn More
WTW™ DURAN™ 500mL Measuring Bottle
Description
- Tested for respective applications
- Supports accurate measurements
- Manufactured from durable DURAN glass
Specifications
Specifications
| Description | 500mL |
| Type | Measuring Bottle |
| For Use With (Application) | Respirometric measurements, soil respiration, degradability tests, biogas examinations |
| Material | DURAN™ Glass |