Learn More
DWK Life Sciences DURAN™ Blank for lid, flat flange, tall form, ground
Shop All DWK Life Sciences ProductsDescription
DURAN™ blanks for flat flange covers can be further processed into customer-specific articles using further DURAN™ components (threaded tubes, standard ground joints, threaded sleeves, etc.). Due to their geometry and solid wall thickness, DURAN™ flat flange covers are pressure and vacuum resistant. For pressure resistance, please refer to our notes in the technical appendix of the laboratory glassware catalogue. For glass articles which deviate from the original form due to further processing, the permissible operating data must be determined in accordance with the technical rules and regulations.
- High form, ground
- Due to the geometry and the massive wall thickness, the blanks are suitable for devices with pressure and vacuum applications.
- Glass articles for further processing
Specifications
Specifications
| Description | DN 120 |
| Material | Borosilicate Glass (3.3) |
| For Use With (Application) | For customer-specific further processing |
| No. per Pack | 4/Pk. |
| Autoclavable | Autoclavable |