Learn More
Lenz Laborglasinstrumente™ DURAN™ Glass Round Bottom Flasks
Can be used to make solutions or hold, contain, collect and measure chemicals. Lenz Laborglasinstrumente™ DURAN™ Glass Round Bottom Flasks are manufactured from durable borosilicate glass.
Brand: Lenz Laborglasinstrumente™ B0418351
Additional Details : Weight : 18.00000kg
Description
- With inscription label
- Available in a range of capacities
- Corrosion-free and resistant to thermal shock
Specifications
500 mL | |
Borosilicate Glass, Duran Glass | |
Head Touches NSH 29/32 | |
Round |