Learn More
Karl Hecht™ Assistent™ DURAN™ Round Bottom Flasks
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Closure Size:
29/32
NS 14/23
NS 19/26
NS 29/32
Unit Size:
Each
Description
- Without stopper
- With standard ground
- Reinforced ground neck marked with standard ground size
Specifications
Specifications
| Capacity (Metric) | 100 mL |
| Material | Borosilicate Glass, Duran Glass |
| Bottom Shape | Round |
| Closure Size | NS 14/23 |