Learn More
DWK Life Sciences DURAN™ Capillary-Single way Stopcock, complete with SBW-J-key
Shop All DWK Life Sciences Products
Click to view available options
Plug Bore:
2 mm
2.5 mm
Closure Size:
12.5 NS
14.5 NS
Unit Size:
Pack of 50
Description
DURAN™ single way stopcocks complete with threaded valve plugs are the most common form of ground shut-off taps. The tap plug is secured in the tap sleeve by a 3-part tap safety catch.
- Ground shut-off valve sleeve with SBW-J valve plug
- The 3-part tap safety catch secures the tap plug in the tap sleeve
- Taper 1:10
- Glass articles for further processing
Especificaciones
Especificaciones
| Plug Bore | 2 mm |
| Diameter (Metric) | 8 mm |
| Material | Borosilicate Glass (3.3), SBW-J, Polypropylene |
| Autoclavable | Not Autoclavable |
| Closure Size | 12.5 NS |
| No. per Pack | 50/Pk. |
| For Use With (Application) | Installation in glass apparatus and devices |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido