Learn More
Hirschmann™ DURAN™ Class AS Automatic Burettes White Graduations
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
Includes:
2000 mL Brown Glass Storage Bottle
No bottle
Unit Size:
Each
Description
- Pellet
- With white graduation
- Amber glass
- Acc. to DIN EN ISO 385
Specifications
Specifications
| Color | Brown |
| Capacity (Metric) | 50 mL |
| Length (Metric) | 500 mm |
| Material | Glass |
| Graduations | 0.1 mL |
| Includes | No bottle |
| Scale | White |
| Stopcock | Lateral |
| Stopcock Material | PTFE |
| Tolerance | ±0.05 mL |
| Show More |