Learn More
Hirschmann™ DURAN™ Class AS Automatic Burettes White Graduations
Product Code. 10606911
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
Includes:
2000 mL Brown Glass Storage Bottle
No bottle
Unit Size:
Each
Product Code. 10606911
Brand: Hirschmann™ 3550375
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Excellent for sensitively titrating amounts of chemical solutions in the laboratory
  • Pellet
  • With white graduation
  • Amber glass
  • Acc. to DIN EN ISO 385
Specifications
Color Brown
Capacity (Metric) 50 mL
Length (Metric) 500 mm
Material Glass
Graduations 0.1 mL
Includes 2000 mL Brown Glass Storage Bottle
Scale White
Stopcock Lateral
Stopcock Material PTFE
Tolerance ±0.05 mL
Type Automatic Titrating Buret
Class Class AS
Certifications/Compliance DIN EN ISO 385