Learn More
Lenz Laborglasinstrumente™ Erlenmeyer Flasks With Conical Ground Joint
Product Code. 12034098
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
200 mL
250 mL
300 mL
500 mL
1000 mL
Standard Taper Joint:
NS19/26
NS29/32
Unit Size:
Each
Pack of 10
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Standard Taper Joint Unit Size
12034098 100 mL NS19/26 Pack of 10
15340895 1000 mL NS29/32 Pack of 10
10298070 100 mL NS19/26 Each
10662351 100 mL NS29/32 Pack of 10
15390885 200 mL NS29/32 Pack of 10
10741434 250 mL NS29/32 Pack of 10
10017750 300 mL NS29/32 Each
11965976 300 mL NS29/32 Pack of 10
10434061 500 mL NS29/32 Pack of 10
10533831 50 mL NS19/26 Pack of 10
10652351 50 mL NS29/32 Each
12163991 50 mL NS29/32 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 12034098 Supplier Lenz Laborglasinstrumente™ Supplier No. 03021937
  • ONLINE EXCLUSIVE PRICE
    € 139.00
    € 84.32 / Pack of 10
    Save € 54.68
    39% Off
View all promotions
Estimated Shipment: 05-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For the safe and easy mixing, pouring and storage of chemicals in the laboratory. Lenz Laborglasinstrumente™ Erlenmeyer Flasks with Conical Ground Joint are manufactured from durable DURAN™ borosilicate glass.
  • Acc. to DIN EN ISO 4797
  • With scale (approx. volume/mL)
  • With inscription label
  • Corrosion-free and resistant to thermal shock
  • With conical joint, featuring high taper accuracy and excellent tightness
Specifications
Certifications/Compliance DIN EN ISO 4797
Capacity (Metric) 100 mL
Material Borosilicate Glass, Duran Glass
Includes With scale (approx. volume/mL), with inscription label
Standard Taper Joint NS19/26
Diameter (Metric) Outer 64 mm