Learn More
Lenz Laborglasinstrumente™ DURAN™ Four-neck Round Bottom Flasks with Angled Side Necks 7°
Specifications
Specifications
| Certifications/Compliance | Acc. to DIN 12392 and in addition to DIN standard |
| Capacity (Metric) | 250 mL |
| Material | Borosilicate Glass, Duran Glass |
| Socket Joint | 29/32 NS Side Neck, 29/32 NS Center Neck |
| Bottom Shape | Round |