Learn More
DWK Life Sciences DURAN™ Funnel, with short stem
Product Code. 10758551 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric)Top:
35 mm
45 mm
55 mm
70 mm
80 mm
100 mm
120 mm
150 mm
180 mm
200 mm
300 mm
Unit Size:
Each
Pack of 10
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric)Top Unit Size
10758551 180 mm Each
11772352 55 mm Pack of 10
11782352 80 mm Pack of 10
11792352 100 mm Pack of 10
11702362 150 mm Pack of 10
10455651 200 mm Each
10323721 300 mm Each
11736153 45 mm Pack of 10
11746153 70 mm Pack of 10
13197164 35 mm Pack of 10
11756153 120 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 10758551 Supplier DWK Life Sciences Supplier No. 213515905
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A funnel enables a clean transfer and filtration of liquids.
Transferring liquids and solids is a common working method. In this activity the funnel is used, which due to its shape can absorb a relatively large amount of liquid or solid and transfer it through a constriction into a vessel with a small opening. Funnels are available in different sizes and designs, selected according to their application.
  • With short stem
  • Due to its shape, it is suitable for liquid-solid filtration using a filter paper
  • Fired-on, highly durable white print
Specifications
Diameter (Metric)Top 180 mm
Material Borosilicate Glass (3.3)
Autoclavable Autoclavable
For Use With (Application) Decanting and filtration of liquids; The spillage of substances is avoided; Filtering
Quantity 1 Ea.
Product Type Non-DIN/ISO size