Learn More
Lenz Laborglasinstrumente™ Duran™ Glass Screw Thread Tube
Click to view available options
Cone Size:
NS 14/23
NS 19/26
NS 29/32
Thread Size:
GL 14
GL 18
GL 25
Unit Size:
Each
Description
- Duran™ Glass
Specifications
Specifications
| Cone Size | NS 29/32 |
| Includes | Screw-cap aperture, Silicone sealing ring, PTFE washers |
| Material | Duran Glass |
| Product Type | Screw thread Tube |
| Quantity | 1 Ea. |
| Thread Size | GL 25 |