Learn More
Witeg™ Duran Glass Separatory Funnels
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
1000 mL
2000 mL
Unit Size:
Each
Description
- Conical shape
- Graduated
- Economy-needle valve stopcock
- PTFE-needle valve
- Screw-thread and cap
Specifications
Specifications
| Capacity (Metric) | 100 mL |
| Material | Duran Glass, Borosilicate Glass |
| Quantity | 1Pack |
| Product Type | Separatory Funnel |