Learn More
Lenz Laborglasinstrumente™ DURAN™ Glass Stirrer
Product Code. p-8006044
Click to view available options
For Use With (Equipment):
Flask, 10,000 mL
Flask, 100 mL
Flask, 1000 mL
Flask, 2000 mL
Flask, 250 mL
Flask, 250/500 mL
Flask, 250/500 mL, 10 GL Thread
Flask, 4000/5000 mL
missing translation for 'unitSize'
Each
Product Code. 11576834
missing translation for 'mfr': Lenz Laborglasinstrumente™ 06307001
Glass Stirrer made of Duran™ glass.
  • Duran™ Glass
Specifications
For Use With (Equipment) Flask, 100 mL
Product Type Stirrer
Material Glass