Learn More
DWK Life Sciences DURAN™ Glass Stopper, Clear, Standard Taper Ground Joint, Octagonal flat head
Clear DURAN™ glass ground stopper.
€ 65.70 - € 114.00
Specifications
Autoclavable | No |
---|---|
Material | Boro 3.3 |
For Use With (Application) | Storage of air sensitive reagents, and chemicals; Storage of hygroscopic solids, granules and powders; Storage of pharmaceutical APIs; Sealing laboratory glassware such as flasks, reagent bottles, and mixing cylinders |
Quantity | 10/Pk. |
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
15251779
|
DWK Life Sciences
216240401 |
ST 12/21 |
€ 65.70
Pack of 10 |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||||
15814508
|
DWK Life Sciences
216240307 |
ST 10/19 |
€ 114.00
Pack of 10 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
15425924
|
DWK Life Sciences
216240701 |
ST 19/26 |
€ 94.50
Pack of 10 |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
15553162
|
DWK Life Sciences
216240804 |
ST 24/29 |
€ 99.80
Pack of 10 |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||||
Description
Semi-hollow or solid DURAN™ glass stoppers can be used to close glassware items such as flasks, separatory funnels or reagent bottles with ground joint necks. The Standard Taper ground glass joint forms a tight, leak-proof seal. Use grease for heating or vacuum applications. With octagonal shaped head. Note on using DURAN™ glass stoppers with soda-lime glass bottles: If the bottle and the stopper have a temperature difference greater than 30°C, the glass stoppers can become stuck!
- All DURAN™ glass components and therefore suitable for storage of aggressive materials
- Tight closure due to Standard Taper ground joint
- Octagonal shaped head
Specifications
No | |
Boro 3.3 | |
Storage of air sensitive reagents, and chemicals; Storage of hygroscopic solids, granules and powders; Storage of pharmaceutical APIs; Sealing laboratory glassware such as flasks, reagent bottles, and mixing cylinders | |
10/Pk. |