Learn More
Lenz Laborglasinstrumente™ Tubes with Socket and Cone
Provides for quick and easy leak-tight connection of apparatus in the laboratory. Lenz Laborglasinstrumente™ Tubes with Socket and Cone are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 01101422
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10671892
  • € 11.08 / Each
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
Description
Description
  • Available in a range of socket/cone sizes
  • Corrosion-free and resistant to thermal shock
  • With conical joint, featurig high taper accuracy and excellent tightness
Specifications
Specifications
1Pack
Socket Size/Cone Size: NS 19/26