Learn More
Karl Hecht™ DURAN™ Ground Joint Erlenmeyer Flasks
Product Code. 10125861
Change view
Click to view available options
Capacity (Metric):
50 mL
250 mL
2000 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10125861 2000 mL Each
10788652 50 mL Each
10748822 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10125861 Supplier Karl Hecht™ Supplier No. 41915056
Provided with a reinforced standard ground neck with joint size clearly and durably marked.
  • Graduated with durable white markings
  • Blank label area included
  • Manufactured from DURAN™ borosilicate glass 3.3
  • Stoppers sold separately
Specifications
Color Clear
Capacity (Metric) 2000 mL
Material Borosilicate Glass (3.3)
Neck Style Reinforced
Standard Taper Joint 29/32
Writing Surface White
Graduated Yes
Bottom Shape Flat