Learn More
DWK Life Sciences DURAN™ Hexagonal Stopper, short ground, flat, finished
Product Code. 15981521 Shop All DWK Life Sciences Products
Click to view available options
Size:
NS 10/13
NS 12/14
NS 14/15
NS 19/17
NS 24/20
NS 29/22
Unit Size:
Pack of 100
Product Code. 15981521
Brand: DWK Life Sciences 286125004
Only null left
Add to basket
Add to basket
Hollow glass stopper for sealing.
DURAN™ hexagonal hollow plugs as a short ground version suitable for DURAN™ short ground sleeves. Cut quality: Ground.
  • Short grinding
  • Flat
  • Ground
  • Hollow glass stopper
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Closure of vessels with standard ground joints; Used for closing short ground sockets
Quantity 100/Pk.
Size NS 14/15
Autoclavable Autoclavable
Material Borosilicate Glass