Learn More
DWK Life Sciences DURAN™ Hexagonal Stopper, short ground, flat, finished
Shop All DWK Life Sciences Products
Click to view available options
Size:
NS 10/13
NS 12/14
NS 14/15
NS 19/17
NS 24/20
NS 29/22
Unit Size:
Pack of 100
Description
DURAN™ hexagonal hollow plugs as a short ground version suitable for DURAN™ short ground sleeves. Cut quality: Ground.
- Short grinding
- Flat
- Ground
- Hollow glass stopper
Specifications
Specifications
| For Use With (Application) | Closure of vessels with standard ground joints; Used for closing short ground sockets |
| Quantity | 100/Pk. |
| Size | NS 14/15 |
| Autoclavable | Autoclavable |
| Material | Borosilicate Glass |