missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Microbiology Bottle, Narrow Neck, Unground

Product Code. 11699848 Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
20,000 mL
2000 mL
25 mL
250 mL
50 mL
500 mL
5000 mL
Unit Size:
Each
Pack of 10
This item is not returnable. View return policy

Product Code. 11699848

Brand: DWK Life Sciences 211612403

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Culture bottles with a narrow neck for use with rubber stoppers

The culture bottles are moulded from DURAN™ borosilicate 3.3 Type 1 glass, and are suitable for the preparation of media or culturing of microorganisms. The bottles comply with ISO 4796-2 and feature a narrow conical unground neck for use with rubber stoppers. Suitable for autoclaving at 121 or 134°C or dry heat sterilization.

  • Narrow neck for rubber stoppers
  • Retrace code
  • Autoclavable at 121°C or 134°C
  • Supplied without stopper
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 100 mL
For Use With (Application) Autoclaving of media and reagents; Preparation of media and reagents; Culturing vessel
Includes With print
Material Borosilicate Glass (3.3)
Quantity 10/Pk.
Diameter (Metric) 52 mm
Autoclavable Autoclavable
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.