Learn More
DWK Life Sciences DURAN™ Separating Funnel stopcock Gilson, complete with PTFE key
Shop All DWK Life Sciences Products
Click to view available options
Closure Size:
12.5 NS
14.5 NS
18.8 NS
Unit Size:
Pack of 25
Description
DURAN™ Gilson separating funnel taps are components for the production of Gilson separating funnels with a particularly strong approach to reducing the risk of breakage. In the polished version with PTFE plug suitable for grease free working.
- Separating funnel tap sleeve with PTFE plug
- Fat-free working through the polished tap surface
- With a particularly strong approach to reducing the risk of breakage
- Glass articles for further processing
Specifications
Specifications
| Plug Bore | 2 mm |
| Diameter (Metric) | 16 mm |
| Material | Borosilicate Glass (3.3), PTFE |
| Autoclavable | Not Autoclavable |
| Closure Size | 12.5 NS |
| Width (Metric) | 1.8 mm |
| No. per Pack | 25/Pk. |
| For Use With (Application) | For attachment to DURAN™ separating funnel raw bodies; For the production of separating funnels according to Gilson |