Learn More
DWK Life Sciences DURAN™ Separating Funnel Stopcock with short inlet, complete with PTFE key
Product Code. 15804758 Shop All DWK Life Sciences Products
Click to view available options
Closure Size:
12.5 NS
14.5 NS
18.8 NS
21.5 NS
Unit Size:
Pack of 25
Product Code. 15804758
Brand: DWK Life Sciences 286433204
Only null left
Add to basket
Add to basket
For easier attachment to DURAN™ separating funnel raw bodies.
DURAN™ Separating funnel taps with short funnel are components for the production of Gilson separating funnels with a particularly strong approach to reducing the risk of breakage with a conical shape right down to the socket of the valve. In the polished version with PTFE plug suitable for grease free working.
  • Separating funnel tap sleeve with PTFE plug
  • with short approach funnel
  • With a particularly strong approach to reducing the risk of breakage
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Plug Bore 6 mm
Material Borosilicate Glass (3.3), PTFE
Autoclavable Autoclavable
Closure Size 21.5 NS
No. per Pack 25/Pk.
For Use With (Application) For attachment to DURAN™ separating funnel raw bodies