Learn More
DWK Life Sciences DURAN™ Separating Funnel Stopcock with short inlet, complete with PTFE key
Shop All DWK Life Sciences Products
Click to view available options
Closure Size:
12.5 NS
14.5 NS
18.8 NS
21.5 NS
Unit Size:
Pack of 25
Description
DURAN™ Separating funnel taps with short funnel are components for the production of Gilson separating funnels with a particularly strong approach to reducing the risk of breakage with a conical shape right down to the socket of the valve. In the polished version with PTFE plug suitable for grease free working.
- Separating funnel tap sleeve with PTFE plug
- with short approach funnel
- With a particularly strong approach to reducing the risk of breakage
- Glass articles for further processing
Specifications
Specifications
| Plug Bore | 2.5 mm |
| Material | Borosilicate Glass (3.3), PTFE |
| Autoclavable | Autoclavable |
| Closure Size | 14.5 NS |
| No. per Pack | 25/Pk. |
| For Use With (Application) | For attachment to DURAN™ separating funnel raw bodies |