Learn More
DWK Life Sciences DURAN™ Single way Stopcock, complete with hollow key with glass handle
Shop All DWK Life Sciences Products
Click to view available options
Closure Size:
12.5 NS
14.5 NS
18.8 NS
21.5 NS
24 NS
29.2 NS
Unit Size:
Each
Pack of 25
Pack of 5
Pack of 50
Description
DURAN™ single way stopcocks complete with hollow DURAN™ cock plug are well suited for work with temperature changes and higher tightness requirements.
- Ground shut-off valve sleeve with hollow glass plug with glass handle
- High tightness
- Taper 1:10
- Glass articles for further processing
Specifications
Specifications
| Plug Bore | 2.5 mm |
| Diameter (Metric) | 9 mm |
| Material | Borosilicate Glass (3.3) |
| Autoclavable | Autoclavable |
| Closure Size | 14.5 NS |
| Width (Metric) | 1.5 mm |
| No. per Pack | 50/Pk. |
| For Use With (Application) | When working with temperature changes and higher tightness requirements; Installation in glass apparatus and devices |