Learn More
DWK Life Sciences DURAN™ Single way Stopcock, ground and polished, without key, Taper 1:5
Product Code. 15894788 Shop All DWK Life Sciences Products
Click to view available options
Plug Bore:
2 mm
2.5 mm
4 mm
6 mm
8 mm
10 mm
Unit Size:
Pack of 25
Pack of 5
Pack of 50
Product Code. 15894788
Brand: DWK Life Sciences 286456409
Only null left
Add to basket
Add to basket
Ground and polished shut-off taps with 1:5 pitch without plug for grease free operation.
Ground and polished DURAN™ single way stopcocks with an alternative pitch of 1:5 are shut-off valves which allow grease free operation with corresponding PTFE valve plugs and are particularly suitable for vacuum applications. The larger taper angle prevents the cock plug from jamming.
  • Valve sleeve ground and polished, without plug
  • Fat-free working through the polished tap surface
  • Taper 1:5
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Plug Bore 6 mm
Diameter (Metric) 13 mm
Material Borosilicate Glass (3.3)
Autoclavable Autoclavable
Closure Size 19.5 NS
Width (Metric) 1.5 mm
No. per Pack 25/Pk.
For Use With (Application) Suitable for installation in glass apparatus for vacuum applications