Learn More
DWK Life Sciences DURAN™ Single way Stopcock, ground and polished, without key, Taper 1:5
Shop All DWK Life Sciences Products
Click to view available options
Plug Bore:
2 mm
2.5 mm
4 mm
6 mm
8 mm
10 mm
Unit Size:
Pack of 25
Pack of 5
Pack of 50
Description
Ground and polished DURAN™ single way stopcocks with an alternative pitch of 1:5 are shut-off valves which allow grease free operation with corresponding PTFE valve plugs and are particularly suitable for vacuum applications. The larger taper angle prevents the cock plug from jamming.
- Valve sleeve ground and polished, without plug
- Fat-free working through the polished tap surface
- Taper 1:5
- Glass articles for further processing
Specifications
Specifications
| Plug Bore | 6 mm |
| Diameter (Metric) | 13 mm |
| Material | Borosilicate Glass (3.3) |
| Autoclavable | Autoclavable |
| Closure Size | 19.5 NS |
| Width (Metric) | 1.5 mm |
| No. per Pack | 25/Pk. |
| For Use With (Application) | Suitable for installation in glass apparatus for vacuum applications |