Learn More
DWK Life Sciences DURAN™ Stainless Steel Multiport GL 45 Connector Cap, from PBT, GL 45, with 2 Ports
Shop All DWK Life Sciences ProductsDescription
- Robust and durable tubing connection system compatible with all sizes of DURAN™ GL 45 laboratory and media bottles
- Facilitates transfer of liquids within a closed and sterile system via the bottle top adaptor
- Two barbed male hose connectors suitable for flexible silicone tubing with 8.0 mm inner diameter
- Freely rotating stainless steel centre allows the bottle to be unscrewed without disconnecting the tubing
- Connectors and body manufactured from Type 316L (1.4404 / S31603) stainless steel
- Hygienic, electropolished surface for efficient cleaning and sterilisation
- All components fully welded to avoid entrapment voids
- USP <88> Class VI silicone gasket and red PBT GL 45 screw cap provide a liquid-tight seal
- Wide temperature usage range up to 180°C; autoclavable at 121°C for 15 minutes
- Lot coded for full traceability
- Ideal for chemistry, life science and biopharma laboratories
Specifications
Specifications
| Color | Red |
| For Use With (Application) | Sterile fluid transfer in and out of lab bottles; Supply of reagents to instruments; Port assemblies for spinner flasks, and cell culture vessels; Autoclaving of media in bottles |
| Material | Stainless Steel (316L) |
| Quantity | Case of 1 Connector Cap |
| Sterility | Non-sterile |
| Thread Size (Metric) | 92 mm |
| Neck Finish | 92 mm |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Product Type | Connector Cap (2-port) |