Learn More
DWK Life Sciences DURAN™ Stopcock Key, ground, with groove
Product Code. p-7189483 Shop All DWK Life Sciences Products
Click to view available options
Closure Size:
10.0 NS
21.5 NS
24.0 NS
29.2 NS
Unit Size:
Pack of 25
Pack of 50
Product Code. 15844518
Brand: DWK Life Sciences 286036209
Only null left
Add to basket
Add to basket
For use with ground DURAN™ single way stopcocks.
DURAN™ tap plugs with groove are solid tap plugs which are secured by an O-ring.
  • With groove, polished
  • The groove serves to hold the O-ring, which secures the plug from falling out of the tap.
  • Solid glass tap plugs
  • Taper 1:10
TRUSTED_SUSTAINABILITY
Specifications
Plug Bore 8.0 mm
Material Borosilicate Glass (3.3)
Autoclavable Autoclavable
Closure Size 24.0 NS
No. per Pack 25/Pk.