Learn More
DWK Life Sciences DURAN™ Stopcock Key, hollow, threaded glass handle, ground, with three part retaining device
To be used for higher tightness requirements.
€ 630.00 - € 1675.00
Specifications
Material | Borosilicate Glass (3.3), Polypropylene |
---|---|
Autoclavable | Yes |
For Use With (Application) | When working with temperature changes and higher tightness requirements |
Product Code | Brand | Closure Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Size | Price | Quantity & Availability | |||||
15941511
|
DWK Life Sciences
286046201 |
21.5 NS |
€ 970.00
Pack of 25 |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
15814538
|
DWK Life Sciences
286047009 |
24 NS |
€ 1320.00
Pack of 25 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
15884528
|
DWK Life Sciences
286044406 |
18.8 NS |
€ 630.00
Pack of 25 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
15864528
|
DWK Life Sciences
286043007 |
14.5 NS |
€ 1030.00
Pack of 50 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
15854528
|
DWK Life Sciences
286041803 |
12.5 NS |
€ 975.00
Pack of 50 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
15824538
|
DWK Life Sciences
286047206 |
29.2 NS |
€ 1675.00
Pack of 25 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Description
Hollow DURAN™ tap plugs are well suited for work with temperature changes and higher tightness requirements.
- Hollow tap plug with thread, glass handle and 3-piece tap safety device
Specifications
Borosilicate Glass (3.3), Polypropylene | |
Yes | |
When working with temperature changes and higher tightness requirements |