Learn More
DWK Life Sciences DURAN™ Tube with Screw Thread, with DIN thread
The threaded tube is used for further processing in glass components.
€ 34.45 - € 105.00
Specifications
Autoclavable | Yes |
---|---|
Material | Boro 3.3 |
For Use With (Application) | Shoring in glass apparatuses |
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
11742919
|
DWK Life Sciences
248360207 |
GL 14 |
€ 36.95
Pack of 10 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
11752919
|
DWK Life Sciences
248370105 |
GL 18 |
€ 43.15
Pack of 10 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
13167613
|
DWK Life Sciences
248350103 |
GL 45 |
€ 34.45
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
11762919
|
DWK Life Sciences
248380209 |
GL 25 |
€ 56.30
Pack of 10 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
11772919
|
DWK Life Sciences
248390107 |
GL 32 |
€ 105.00
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
Description
This threaded tube has a screw thread and can be incorporated into glass components by a further processing step.
- With DIN thread
Specifications
Yes | |
Boro 3.3 | |
Shoring in glass apparatuses |