Learn More
DWK Life Sciences DURAN™ Tube with two Hose Connections
Product Code. 15874508 Shop All DWK Life Sciences Products
missing translation for 'orderingAttributeHoverText'
Joint Size:
6 mm
7 mm
8 mm
9 mm
10 mm
11 mm
12 mm
14 mm
missing translation for 'unitSize'
Pack of 125
Pack of 250
Pack of 500
Product Code. 15874508
missing translation for 'mfr': DWK Life Sciences 286021604
Only null left
Add to basket
Add to basket
For attaching to a wide variety of apparatus.
Hoses can be connected to laboratory apparatus via DURAN™ tube nozzles. There is always a risk of injury due to glass breakage when dismantling tightly fitted hoses from glass olives. DWK therefore recommends the use of DURAN™ glass threads with plastic nozzles.
  • Hose olives enable the connection of hoses to laboratory apparatuses
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Quantity 500/Pk.
Item Description For attaching to various apparatuses; Installation in different apparatus possible
Type Tube with Connections
Joint Size 7 mm