Learn More
DWK Life Sciences DURAN™ Weighing Bottle Tops, unground
Product Code. 15884848 Shop All DWK Life Sciences Products
Click to view available options
Closure Size:
NS 19
NS 24
NS 29
NS 34
NS 40
NS 50
NS 60
NS 80
Unit Size:
Pack of 240
Pack of 250
Pack of 90
Product Code. 15884848
Brand: DWK Life Sciences 286605404
Only null left
Add to basket
Add to basket
Weighing glass lid for grinding and completing.
DURAN™ weighing bottles are used for weighing chemicals on analysis trolleys. The tight seal of the replaceable, ground weighing glass lid prevents sample loss during transport after weighing.
  • Unground
  • Subsequent grinding results in a tight seal of the weighing lid in the weighing pan. A loss of sample can thus be avoided.
  • Grinding glass product
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Weighing of chemicals
Material Borosilicate Glass (3.3)
Quantity 250/Pk.
Closure Size NS 40