Learn More
DWK Life Sciences DURAN™ Weighing Bottle Tops, unground
Shop All DWK Life Sciences Products
Click to view available options
Closure Size:
NS 19
NS 24
NS 29
NS 34
NS 40
NS 50
NS 60
NS 80
Unit Size:
Pack of 240
Pack of 250
Pack of 90
Description
DURAN™ weighing bottles are used for weighing chemicals on analysis trolleys. The tight seal of the replaceable, ground weighing glass lid prevents sample loss during transport after weighing.
- Unground
- Subsequent grinding results in a tight seal of the weighing lid in the weighing pan. A loss of sample can thus be avoided.
- Grinding glass product
Specifications
Specifications
| For Use With (Application) | Weighing of chemicals |
| Material | Borosilicate Glass (3.3) |
| Quantity | 250/Pk. |
| Closure Size | NS 40 |
| Autoclavable | Autoclavable |